Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Baboon Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.50 - $670.50
Specifications
| Antigen | Baboon |
|---|---|
| Concentration | 0.75 mg/mL |
| Dilution | Western Blot 1:200, Immunoprecipitation 1:200 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018168
![]() |
Novus Biologicals
NB3006650.025MG |
0.025 mg |
Each for $243.50
|
|
|||||
NB300665005
![]() |
Novus Biologicals
NB3006650.05MG |
0.05 mg |
Each for $402.00
|
|
|||||
NB300665
![]() |
Novus Biologicals
NB300665 |
0.1 mg |
Each for $670.50
|
|
|||||
Description
Baboon Polyclonal specifically detects Baboon in Drosophila samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Baboon | |
| Western Blot 1:200, Immunoprecipitation 1:200 | |
| Polyclonal | |
| Rabbit | |
| Activin A receptor 45A, ATR 1, Atr 45A, Atr I, ATR1, Atr45A, AtrI, Bab, Babo, CG8224, G protein coupled receptor kinase 3, Gprk 3, Gprk3, l(2)k16912, RE55648p, STK E, STKE | |
| IgG | |
| The is unique to Baboon protein. |
| 0.75 mg/mL | |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Drosophila | |
| Synthetic peptide: QPDPSIEEMKKVVCIEKCRPN, corresponding to amino acids 552 to 572 of Baboon.The was generated against a peptide sequence near the C terminal end of the protein.The peptides was amidated for conjugation. | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title