Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE-1 Antibody (137612), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB295479 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB295479 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB295479 Supplier Novus Biologicals Supplier No. FAB931S

Mouse Monoclonal Antibody

BACE-1 Monoclonal antibody specifically detects BACE-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF

Specifications

Antigen BACE-1
Applications Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 137612
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BACE-1, Thr22-Tyr460, Accession # P56817
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23621
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.