Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE-1 Antibody (7G3), Novus Biologicals™
SDP

Catalog No. NB298009 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298009 100 μL
NB298008 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298009 Supplier Novus Biologicals Supplier No. NBP326484100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BACE-1 Monoclonal antibody specifically detects BACE-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen BACE-1
Applications Western Blot, ELISA
Classification Monoclonal
Clone 7G3
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias APP beta-secretase, ASP2, Aspartyl protease 2, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human BACE-1 [UniProt P56817]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 23621
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.