Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE-2 Antibody (1/ 9), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB015475 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015475 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015475 Supplier Novus Biologicals Supplier No. NBP281075UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BACE-2 Monoclonal specifically detects BACE-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BACE-2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 43839
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias AEPLC, Asp 1, ASP1, ASP21, Aspartic-like protease 56 kDa, Aspartyl protease 1, beta secretase 2, beta-secretase 2, beta-site amyloid beta A4 precursor protein-cleaving enzyme 2, Beta-site amyloid precursor protein cleaving enzyme 2, Beta-site APP cleaving enzyme 2, beta-site APP-cleaving enzyme 2, beta-site APP-cleaving enzyme 2, EC 3.4.2310ALP5656 kDa aspartic-like protease, CEAP1, Down region aspartic protease, Down syndrome region aspartic protease, DRAPBAE2, EC 3.4.23, EC 3.4.23.45, Memapsin-1, Membrane-associated aspartic protease 1, Theta-secretase, transmembrane aspartic proteinase Asp1
Gene Symbols BACE2
Host Species Mouse
Immunogen Human BACE-2 244-333.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25825
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.