Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACE-2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177308 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177308 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177308 Supplier Novus Biologicals Supplier No. NBP177308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BACE-2 Polyclonal specifically detects BACE-2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen BACE-2
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2 μg/mL, Immunohistochemistry-Paraffin 2 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. AAF17078
Gene Alias AEPLC, Asp 1, ASP1, ASP21, Aspartic-like protease 56 kDa, Aspartyl protease 1, beta secretase 2, beta-secretase 2, beta-site amyloid beta A4 precursor protein-cleaving enzyme 2, Beta-site amyloid precursor protein cleaving enzyme 2, Beta-site APP cleaving enzyme 2, beta-site APP-cleaving enzyme 2, beta-site APP-cleaving enzyme 2, EC 3.4.2310ALP5656 kDa aspartic-like protease, CEAP1, Down region aspartic protease, Down syndrome region aspartic protease, DRAPBAE2, EC 3.4.23, EC 3.4.23.45, Memapsin-1, Membrane-associated aspartic protease 1, Theta-secretase, transmembrane aspartic proteinase Asp1
Gene Symbols BACE2
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human BACE2. The immunogen is located within the last 50 amino acids of BACE2. Amino Acid Squence: CQRRPRDPEVVNDESS
Molecular Weight of Antigen 57 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 25825
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.