Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BACH1 Antibody (OTI4E11), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP270240FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270240FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270240FR Supplier Novus Biologicals Supplier No. NBP270240FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BACH1 Monoclonal specifically detects BACH1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen BACH1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI4E11
Conjugate DyLight 680
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias basic region leucine zipper transcriptional regulator BACH1, BTB and CNC homolog 1, BTB and CNC homology 1, basic leucine zipper transcription factor 1, HA2303, transcription regulator protein BACH1, transcription regulator protein10BACH-1
Gene Symbols BACH1
Host Species Mouse
Immunogen Full length recombinant protein of human BACH1 (NP_996749) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 571.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.