Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BACH2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584376
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA584376 100 μL
1 options

Catalog No. PIPA584376

Supplier: Invitrogen™ PA584376

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SNSLKPGLAR GQIKSEPPSE ENEEESITLC LSGDEPDAKD RAGDVEMDRK QPSPAPTPTA PAGAACLERS RSVASPSCLR SLFSITK Highest antigen sequence indentity to the following orthologs: Rat - 80%, Mouse - 85%.

BTB and CNC homolog 2 (BACH2) is a transcriptional regulator that acts as repressor or activator. BACH2 binds to Maf recognition elements (MARE). It induces apoptosis in response to oxidative stress through repression of the antiapoptotic factor HMOX1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BACH2
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BACH2
Gene Accession No. Q9BYV9
Gene Alias Bach2; BTB and CNC homolog 2; BTB and CNC homology 1, basic leucine zipper transcription factor 2; BTB and CNC homology 2; BTB domain and CNC homolog 2; BTBD25; E030004N02Rik; RGD1562865; similar to BTB and CNC homology 1, basic leucine zipper transcription factor 2; transcription regulator protein BACH2; Transcription regulator protein BACH2 (BTB and CNC homolog 2)
Gene Symbols BACH2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BACH2. Recombinant protein control fragment (Product #RP-106850).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60468
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.