Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baculovirus Envelope gp64 protein Antibody (AcV1), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP200208X Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP200208X 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP200208X Supplier Novus Biologicals Supplier No. NBP200208AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Baculovirus Envelope gp64 protein Monoclonal specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Flow Cytometry.

Specifications

Antigen Baculovirus Envelope gp64 protein
Applications Flow Cytometry
Classification Monoclonal
Clone AcV1
Conjugate Alexa Fluor 488
Dilution Flow Cytometry
Gene Alias ACNVgp129, AcOrf-128
Host Species Mouse
Immunogen The immunogen for this antibody is baculovirus envelope gp64 protein.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.