Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Baculovirus Envelope gp64 protein Antibody (AcV1), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP20208A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20208A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20208A532 Supplier Novus Biologicals Supplier No. NBP200208AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Baculovirus Envelope gp64 protein Monoclonal specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Flow Cytometry.

Specifications

Antigen Baculovirus Envelope gp64 protein
Applications Flow Cytometry
Classification Monoclonal
Clone AcV1
Conjugate Alexa Fluor 532
Dilution Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ACNVgp129, AcOrf-128
Host Species Mouse
Immunogen The immunogen for this antibody is baculovirus envelope gp64 protein.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.