Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bad Antibody - Mouse Adsorbed, Novus Biologicals™
SDP

Catalog No. NB10056078T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056078T 0.025 mL
NB10056078 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056078T Supplier Novus Biologicals Supplier No. NB10056078SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Bad Polyclonal specifically detects Bad in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Bad
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000
Formulation Whole antisera with 0.02% Sodium Azide
Gene Alias BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter
Gene Symbols BAD
Host Species Rabbit
Immunogen A synthetic peptide corrsponding to amino acids 87-103 (GEEPSPFRGRSRSAPPN) of human Bad was used as immunogen; GenBank no. NP_004313.1. Amino acids 88-103 of the peptide immunogen are 93% conserved in mouse and rat Bad.
Purification Method Unpurified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 572
Test Specificity Bad
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Antisera
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.