Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAD-LAMP/LAMP5 Antibody, Novus Biologicals™
SDP

Catalog No. NBP16256520 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP16256520 20 μL
NBP162565 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP16256520 Supplier Novus Biologicals Supplier No. NBP16256520UL

Rabbit Polyclonal Antibody

BAD-LAMP/LAMP5 Polyclonal specifically detects BAD-LAMP/LAMP5 in Human samples. It is validated for Western Blot.

Specifications

Antigen BAD-LAMP/LAMP5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UJQ1
Gene Alias BADLAMP, C20orf103, chromosome 20 open reading frame 103, dJ1119D9.3, LAMP family protein C20orf103, LAMP5, LAMP-5, lysosomal-associated membrane protein family, member 5, UNC-43
Gene Symbols LAMP5
Host Species Rabbit
Immunogen Synthetic peptides corresponding to C20ORF103 The peptide sequence was selected from the middle region of C20ORF103. Peptide sequence CQAQQTISLASSDPQKTVTMILSAVHIQPFDIISDFVFSEEHKCPVDERE.
Molecular Weight of Antigen 28 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24141
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.