Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAF53A Antibody, Novus Biologicals™
SDP

Catalog No. NBP255376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP255376 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP255376 Supplier Novus Biologicals Supplier No. NBP255376

Rabbit Polyclonal Antibody

BAF53A Polyclonal specifically detects BAF53A in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BAF53A
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias actin-like 6A, actin-like protein 6A, actin-related protein 4,53 kDa BRG1-associated factor A, Actin-related protein Baf53a, Actl6, Arp4, ArpNbeta, BAF complex 53 kDa subunit, Baf53a, BAF53AACTL6, BAF53MGC5382, BRG1-associated factor 53A, hArpN beta, INO80 complex subunit K, INO80KARPN-BETA
Gene Symbols ACTL6A
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:TVRAGYAGEDCPKVDFPTAIGMVVERDDGSTLMEIDGDKGKQGGPTYYIDTNALRVPRENMEAISPLKNGMVEDWD
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Extracellular Matrix
Primary or Secondary Primary
Gene ID (Entrez) 86
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.