Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAF57 Rabbit anti-Human, Mouse, Rat, Clone: 9L5F6, Novus Biologicals™
SDP

Catalog No. NB167650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167650 20 μg
NB167649 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB167650 Supplier Novus Biologicals Supplier No. NBP31627920UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BAF57 Monoclonal antibody specifically detects BAF57 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen BAF57
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone 9L5F6
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BAF57BRG1-associated factor 57, chromatin remodeling complex BRG1-associated factor 57, subfamily e, member 1, SWI/SNF related, matrix associated, actin dependent regulator of chromatin, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin e1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatinsubfamily E member 1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 2-100 of human BAF57 (Q969G3).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 6605
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.