Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFF/BLyS/TNFSF13B Antibody (137314), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB305117 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305117 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305117 Supplier Novus Biologicals Supplier No. FAB1241Z
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAFF/BLyS/TNFSF13B Monoclonal specifically detects BAFF/BLyS/TNFSF13B in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen BAFF/BLyS/TNFSF13B
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 137314
Conjugate DyLight 755
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Host Species Mouse
Immunogen E. coli-derived recombinant human BAFF/BLyS/TNFSF13B, Ala81-Leu285, Accession # Q9Y275
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10673
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.