Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFF/BLyS/TNFSF13B Antibody (C257/1638), Novus Biologicals™
SDP

Catalog No. NB25335620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB25335620 20 μg
NB253356100 100 μg
NB253356200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB25335620 Supplier Novus Biologicals Supplier No. NBP25335620UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAFF/BLyS/TNFSF13B Monoclonal specifically detects BAFF/BLyS/TNFSF13B in Human samples. It is validated for ELISA.

Specifications

Antigen BAFF/BLyS/TNFSF13B
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone C257/1638
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Gene Symbols TNFSF13B
Host Species Mouse
Immunogen Purified recombinant human full-length CD257 protein
Molecular Weight of Antigen 31 kDa
Purification Method Protein A purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cell Cycle and Replication, Cytokine Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 10673
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.