Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFFR/TNFRSF13C Antibody (2403C), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB314783 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB314783 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB314783 Supplier Novus Biologicals Supplier No. FAB1162PEATT594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry
Classification Monoclonal
Clone 2403C
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias BAFFR;BAFF-R;BROMIX;CD268;CVID4;prolixin;Tumor necrosis factor receptor superfamily member 13C
Host Species Rabbit
Immunogen Mouse myeloma cell line, NS0-derived human BAFF R/TNFRSF13C, Ser7-Ala71, Accession # Q96RJ3
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 115650
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.