Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFFR/TNFRSF13C Antibody (7J21) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP222005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222005 0.1 mg
NB019285 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP222005 Supplier Novus Biologicals Supplier No. NBP222005

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rat Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 7J21
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:20, ELISA 1:1000-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. Q9D8D0
Gene Alias B cell-activating factor receptor, BAFF receptor, BAFF-R, BAFFRMGC138235, B-cell-activating factor receptor, BLyS receptor 3, BR3, CD268, CD268 antigen, CVID4, tumor necrosis factor receptor superfamily member 13C, tumor necrosis factor receptor superfamily, member 13C
Gene Symbols TNFRSF13C
Host Species Rat
Immunogen Mouse recombinant BAFFR/TNFRSF13C
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 115650
Reconstitution Reconstitute with sterilized PBS to a final concentration of 0.2 mg/ml.
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.