Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFFR/TNFRSF13C Antibody (9B9), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB172930 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB172930 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB172930 Supplier Novus Biologicals Supplier No. NBP280072AF647
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal antibody specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry
Classification Monoclonal
Clone 9B9
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen Y3 cells transfected with mouse BAFFR/TNFRSF13C.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 115650
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.