Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAFFR/TNFRSF13C Antibody (9B9) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB013201 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB013201 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB013201 Supplier Novus Biologicals Supplier No. NBP280073
Add to Cart
Add to Cart

Rat Monoclonal Antibody

BAFFR/TNFRSF13C Monoclonal specifically detects BAFFR/TNFRSF13C in Mouse samples. It is validated for Flow Cytometry, Functional, CyTOF-ready.

Specifications

Antigen BAFFR/TNFRSF13C
Applications Flow Cytometry, Functional Assay, CyTOF
Classification Monoclonal
Clone 9B9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Functional, CyTOF-ready
Formulation PBS
Gene Symbols TNFRSF13C
Host Species Rat
Immunogen Y3 cells transfected with mouse BAFFR/TNFRSF13C.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cytokine Research, Immunology, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 115650
Test Specificity Recognizes mouse BAFFR/TNFRSF13C.
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.