Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG2 Mouse anti-Human, PerCP, Clone: 29000000000, Novus Biologicals™
SDP

Catalog No. NB162141 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162141 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162141 Supplier Novus Biologicals Supplier No. NBP259476PCP
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BAG2 Monoclonal antibody specifically detects BAG2 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen BAG2
Applications Flow Cytometry
Classification Monoclonal
Clone 29000000000
Conjugate PerCP
Formulation PBS
Gene Alias BAG family molecular chaperone regulator 2, BAG-2, BAG-family molecular chaperone regulator-2, Bcl-2-associated athanogene 2, BCL2-associated athanogene 2, dJ417I1.2, dJ417I1.2 (BAG-family molecular chaperone regulator 2), KIAA0576, MGC149462
Host Species Mouse
Immunogen Recombinant human BAG2 (1-211aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 9532
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.