Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG3 Rabbit anti-Human, Mouse, Clone: 8P3W7, Novus Biologicals™
SDP

Catalog No. NB168029 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168029 100 μg
NB168030 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168029 Supplier Novus Biologicals Supplier No. NBP316469100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BAG3 Monoclonal antibody specifically detects BAG3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen BAG3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8P3W7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BAG family molecular chaperone regulator 3, BAG-3, BAG-family molecular chaperone regulator-3, Bcl-2-associated athanogene 3, BCL2-associated athanogene 3, BCL2-binding athanogene 3, Bcl-2-binding protein Bis, BIS, CAIR-1, Docking protein CAIR-1, MGC104307
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 50-150 of human BAG3 (O95817).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 9531
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.