Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI1 Antibody (480912), FITC, Novus Biologicals™
SDP

Catalog No. NB368685 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB368685 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB368685 Supplier Novus Biologicals Supplier No. FAB49691F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAI1 Monoclonal specifically detects BAI1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen BAI1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 480912
Conjugate FITC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human BAI1, Ala31-Thr879, Accession # O14514
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Angiogenesis, Apoptosis, Cancer, GPCR, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 575
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.