Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB11081586G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11081586G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11081586G Supplier Novus Biologicals Supplier No. NB11081586G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAI1 Polyclonal specifically detects BAI1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen BAI1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. O14514
Gene Alias brain-specific angiogenesis inhibitor 1, FLJ41988, GDAIF
Gene Symbols BAI1
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the human BAI1 protein (within residues 1500-1584). [Swiss-Prot# O14514]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 575
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.