Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI2 Antibody (2528B), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB345758 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB345758 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB345758 Supplier Novus Biologicals Supplier No. FAB9338MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BAI2 Monoclonal specifically detects BAI2 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen BAI2
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2528B
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias brain-specific angiogenesis inhibitor 2, Brain-specific angiongenesis inhibitor-2
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-derived human BAI2, Phe21-Asp911, Accession # O60241-3
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 576
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.