Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAI3 Antibody (409611), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB348038 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB348038 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB348038 Supplier Novus Biologicals Supplier No. FAB3965MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAI3 Monoclonal specifically detects BAI3 in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen BAI3
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 409611
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias brain-specific angiogenesis inhibitor 3, dJ91B17.1 (brain-specific angiogenesis inhibitor 3), KIAA0550MGC133100
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human BAI3, Ala25-Thr880, Accession # O60242
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, GPCR
Primary or Secondary Primary
Gene ID (Entrez) 577
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.