Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAIAP2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP215545 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP215545 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP215545 Supplier Novus Biologicals Supplier No. NBP215545
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAIAP2 Polyclonal specifically detects BAIAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen BAIAP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.7mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:20000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9UQB8
Gene Alias BAI1-associated protein 2IRS-58, BAI-associated protein 2, BAP2, brain-specific angiogenesis inhibitor 1-associated protein 2, Fas ligand-associated factor 3, FLAF3, Insulin receptor substrate p53, Insulin receptor substrate p53/p58, Insulin receptor substrate protein of 53 kDa, IRSp53, IRSp53/58, Protein BAP2
Gene Symbols BAIAP2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human BAIAP2. The exact sequence is proprietary.
Molecular Weight of Antigen 61 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10458
Test Specificity Human, Mouse. Expected to cross react based on sequence identity: Rat (98%), Bovine (98%).
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.