Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAIAP2 Antibody (OTI 1D9), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP271603R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271603R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271603R Supplier Novus Biologicals Supplier No. NBP271603R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAIAP2 Monoclonal specifically detects BAIAP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BAIAP2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI 1D9
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BAI1-associated protein 2IRS-58, BAI-associated protein 2, BAP2, brain-specific angiogenesis inhibitor 1-associated protein 2, Fas ligand-associated factor 3, FLAF3, Insulin receptor substrate p53, Insulin receptor substrate p53/p58, Insulin receptor substrate protein of 53 kDa, IRSp53, IRSp53/58, Protein BAP2
Gene Symbols BAIAP2
Host Species Mouse
Immunogen Full length recombinant protein of human BAIAP2 (NP_006331) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10458.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.