Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAIAP2 Mouse anti-Mouse, Human, Clone: CA-A10-5, MilliporeSigma™

Mouse Monoclonal Antibody

$481.79 - $485.79

Specifications

Antigen BAIAP2
Clone CA-A10-5
Applications Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABT132625
View Documents
MilliporeSigma
MABT132625UL
25 μL
Each for $485.79
Only null left
Add to Cart
 
MABT1326100
View Documents
MilliporeSigma
MABT1326100UL
100 μL
Each for $481.79
Only null left
Add to Cart
 
Description

Description

BAIAP2 Monoclonal antibody specifically detects BAIAP2 in Mouse, Human samples. It is validated for Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blotting.
Specifications

Specifications

BAIAP2
Dot Blot, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
Mouse
Mouse, Human
His-tagged full-length human recombinant Brain-specific angiogenesis inhibitor 1-associated protein 2 (BAIAP2)
Protein G purified
CA-A10-5
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Primary
Stable for 1 year at 2-8°C from date of receipt.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.