Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAIAP3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396517 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396517 0.025 mg
NBP241080 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396517 Supplier Novus Biologicals Supplier No. NBP2410800.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BAIAP3 Polyclonal specifically detects BAIAP3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen BAIAP3
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias BAI1-associated protein 3, BAI-associated protein 3, BAP3KIAA0734MGC138334
Gene Symbols BAIAP3
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide from near the amino terminus of human BAP3. The immunogen is located within amino acids 60 - 110 of BAP3. Amino Acid Squence: CRQSPHPGTHTQV
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8938
Test Specificity BAP3 is known to have two isoforms; this BAP3 antibody will recognize only isoform 2. Lower molecular weight bands may represent cleavage or degradation products.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.