Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAK Antibody (38E2), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N174026PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N174026PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N174026PEC5 Supplier Novus Biologicals Supplier No. NBP174026PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAK Monoclonal specifically detects BAK in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BAK
Applications Flow Cytometry
Classification Monoclonal
Clone AT38E2
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Apoptosis regulator BAK, BAK-LIKE, BAKMGC3887, BCL2-antagonist/killer 1, Bcl2-L-7, BCL2L7bcl-2 homologous antagonist/killer, BCL2-like 7 protein, Bcl-2-like protein 7, CDN1, MGC117255, pro-apoptotic protein BAK
Gene Symbols BAK1
Host Species Mouse
Immunogen Recombinant human BAK1 (29-187aa) purified from E.coli
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 578.0
Test Specificity Anti-human BAK1 mAb, clone 8B4, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human BAK1 protein.
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.