Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAMBI/NMA Antibody (OTI5D5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP271778A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP271778A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP271778A32 Supplier Novus Biologicals Supplier No. NBP271778AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAMBI/NMA Monoclonal specifically detects BAMBI/NMA in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen BAMBI/NMA
Applications Western Blot
Classification Monoclonal
Clone OTI5D5
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias BMP and activin membrane-bound inhibitor homolog, BMP and activin membrane-bound inhibitor homolog (Xenopus laevis), NMANon-metastatic gene A protein, Putative transmembrane protein NMA
Gene Symbols BAMBI
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 21-152 of human BAMBI/NMA (NP_036474) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 25805
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.