Learn More
Allegro™ Banana Oil Fit-Test Solutions
Isoamyl Acetate, also referred to as Banana Oil, is a qualitative fit test agent used to detect respirator leakage.
$33.90
Specifications
Product Type Banana Oil Solution
For Use With (Application) Respirator Fit Testing
Quantity 1 Ea.
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Capacity (Metric) Price Quantity & Availability  
19-088-507
View Documents
Allegro™
020301
1 oz. 29.5 mL
Each for $33.90
Only null left
Add to Cart
19-088-505
View Documents
Allegro™
020201
8 oz. 237 mL N/A N/A N/A
Description
Description
  • Banana oil (isoamyl acetate) is the substance originally used for testing respirator fit; its strong, recognizable odor quickly indicates the lack of proper fit
  • Available in two fit test kits and Fit Check Ampules that provide a fast and easy method of on-the-spot qualitative fit testing
Order Info
The Standard Kit comes in a handy carry bag and includes two 1 oz. (30mL) bottles of testing agent, a vinyl test enclosure, measuring pipette, and paper towels. The Deluxe Kit comes in a convenient storage case and contains everything necessary to perform a proper fit test, including an 8 1oz. (30mL) bottles of testing solution, a fit test tent, jars, paper towels, disposable bags, and other necessary components. Replacement banana oil solution is available in 1 oz. (30mL) bottles.
Specifications
Specifications
Banana Oil Solution
Respirator Fit Testing
1 Ea.
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.