Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Band 3 Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB054777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054777 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054777 Supplier Novus Biologicals Supplier No. NBP298703JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Band 3 Polyclonal specifically detects Band 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Band 3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias AE 1, AE1MGC126619, Anion exchange protein 1, Anion exchanger 1, band 3 anion transport protein, BND3, CD233, CD233 antigen, DI, EMPB3, EPB3MGC126623, erythrocyte membrane protein band 3, erythroid anion exchange protein, FR, Froese blood group, MGC116750, MGC116753, RTA1A, Solute carrier family 4 member 1, solute carrier family 4, anion exchanger, member 1 (erythrocyte membraneprotein band 3, Diego blood group), solute carrier family 4, anion exchanger, number 1, SW, Swann blood group, Waldner blood group, WD, WD1, WR, Wright blood group
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human Band 3.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 6521
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.