Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAP1 Antibody (1G8), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB11060521R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11060521R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11060521R Supplier Novus Biologicals Supplier No. NB11060521R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAP1 Monoclonal specifically detects BAP1 in Human samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen BAP1
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 1G8
Conjugate DyLight 550
Dilution Western Blot, Flow Cytometry
Gene Accession No. Q92560
Gene Alias BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Gene Symbols BAP1
Host Species Mouse
Immunogen Full-length human BAP1 protein expressed and purified from E.coli. [UniProt# Q92560]
Molecular Weight of Antigen 85 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8314
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.