Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAP1 Antibody (BAP1/2433), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB386553 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB386553 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB386553 Supplier Novus Biologicals Supplier No. NBP308494AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BAP1 Monoclonal antibody specifically detects BAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen BAP1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BAP1/2433
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Host Species Mouse
Immunogen Recombinant human BAP1 protein fragment (around aa 191-326) (exact sequence is proprietary) (Uniprot: Q92560)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.