Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAP1 Antibody (BAP1/6861R), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB187089 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB187089 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB187089 Supplier Novus Biologicals Supplier No. NBP314042UV
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BAP1 Monoclonal antibody specifically detects BAP1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen BAP1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BAP1/6861R
Conjugate DyLight 350
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase), BRCA1-associated protein 1, Cerebral protein 6, cerebral protein-13, cerebral protein-6, DKFZp686N04275, EC 3.4.19.12, FLJ35406, FLJ37180, hucep-6, KIAA0272HUCEP-13, ubiquitin carboxyl-terminal hydrolase BAP1, ubiquitin carboxy-terminal hydrolase, UCHL2
Host Species Rabbit
Immunogen Recombinant fragment (around aa400-729) of human BAP1 protein (exact sequence is proprietary) (Uniprot: Q92560)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.