Learn More
Barium chloride, anhydrous, 99.998% (metals basis)
Description
Barium chloride is used in manufacture of pigments, fire works, other barium salts, fireworks , hardening of steel, heat treatment salts, purification of brine solution in caustic chlorine plants, lubrication oil additive, textile dye, pigments, white leather, aluminum refining, boiling water treatment and porcelain enamels for sheet steel. It is also used to remove sulfate ion in some electrolytic plants, caustic soda, magnesium metal or sodium metal. It is also used as a component in a flux used to prevent oxidation of molten magnesium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 1560°C |
Melting Point | 963°C |
Color | White |
Physical Form | Lump/Powder |
Quantity | 5 g |
UN Number | UN1564 |
Merck Index | 14,971 |
Solubility Information | Soluble in water and methanol. Insoluble in acids,ethanol,acetone and ethyl acetate. Slightly soluble in nitric acid and hydrochloric acid. |
IUPAC Name | barium(2+);dichloride |
Formula Weight | 208.25 |
Show More |
Safety and Handling
Hazard Category | H301-H332 |
Hazard Statement | GHS H Statement H301-H332 Toxic if swallowed. Harmful if inhaled. |
Precautionary Statement | P261-P264b-P270-P271-P301+P310-P304+P340-P312-P330-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S. |
EINECSNumber | 233-788-1 |
RTECSNumber | CQ8750000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.