Learn More
Barium hydroxide monohydrate, 95%
Description
Barium hydroxide monohydrate is used in water purification. It is used to prepare lubricating and oil additives as well as a precursor to other barium compounds. It is also used to dehydrate and remove sulfate from various products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline |
| Quantity | 500 g |
| Linear Formula | Ba(OH)2·H2O |
| UN Number | UN3262 |
| Solubility Information | Soluble in water. |
| Formula Weight | 189.38 (171.36 Anhydrous) |
| Percent Purity | 95% |
| Odor | Odorless |
| Sensitivity | Air sensitive; Hygroscopic |
| Show More |
Safety and Handling
| Hazard Category | H302+H332-H314 |
| Hazard Statement | GHS H Statement H301-H314-H318-H332 Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if inhaled. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 241-234-5 |
| RTECSNumber | CQ9200000 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.