Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium hydroxide octahydrate, 98+%, for analysis

Catalog No. AC203142500
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle

Catalog No. AC203142500

Supplier: Thermo Scientific Chemicals 203142500

Only null left
Add to Cart

Barium hydroxide octahydrate, 98+%, for analysis, Quantity: 1kg, Packaging: Plastic bottle, CAS: 12230-71-6, White, Grade: Analysis, Melting Point: 78.0 deg.C, Molecular Formula: H2BaO2.8H2O, Molecular Weight: 315.48, Percent Purity: 98+% | CAS: 12230-71-6 | BaH18O10 | 315.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12230-71-6
Molecular Formula BaH18O10
Molecular Weight (g/mol) 315.46
MDL Number MFCD00149152
InChI Key ZUDYPQRUOYEARG-UHFFFAOYSA-L
Synonym barium hydroxide octahydrate, unii-l5q5v03tbn, l5q5v03tbn, dihydroxybarium octahydrate, barium dihydroxide octahydrate, barium hydroxide octahydrate mi, barium hydroxide ba oh 2 , octahydrate, acmc-1an7p, barium hydroxide octa hydrate, barium hydroxide octa-hydrate
PubChem CID 17749109
SMILES O.O.O.O.O.O.O.O.[OH-].[OH-].[Ba++]

Specifications

Melting Point 78.0°C
Color White
Physical Form Crystals, Crystalline Flakes or Powder
Quantity 250 g
Assay Percent Range 98+%
Linear Formula Ba(OH)2·8H2O
Merck Index 15, 973
Solubility Information Solubility in water: 56g/L (15°C). Other solubilities: freely soluble in methanol, slightly soluble in ethanol, practically insoluble in acetone
IUPAC Name barium(2+) octahydrate dihydroxide
Formula Weight 315.48
Percent Purity ≥98%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 2.18
Density 2.1800g/mL
Chemical Name or Material Barium hydroxide octahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.