Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium iodide hydrate, 98%, for analysis

Catalog No. ac208000500
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC208000500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC208000500 Supplier Thermo Scientific Chemicals Supplier No. 208000500
Only null left
Add to Cart

CAS: 85017-89-6 | BaI2 | 391.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85017-89-6
Molecular Formula BaI2
Molecular Weight (g/mol) 391.14
MDL Number MFCD00149158
InChI Key SGUXGJPBTNFBAD-UHFFFAOYSA-L
IUPAC Name barium(2+) diiodide
SMILES [I-].[I-].[Ba++]

Specifications

Melting Point 740.0°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 97.5% min. (on dry substance) (Argentometry)
Linear Formula BaI2·xH2O
Merck Index 14, 980
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol and acetone
Formula Weight 391.14
Percent Purity 98%
Grade Analytical
Packaging Glass bottle
Chemical Name or Material Barium iodide hydrate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not eat,drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physici

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.