Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium zirconium oxide, 99% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4408064
Click to view available options
Quantity:
250 g
1 kg

Barium zirconium oxide is involved in the preparation of its nano powder, which finds application in the gas sensing performance of thick films especially with ammonia gas. Copper(II) oxide doping with yttrium-doped barium zirconate is used as an electrolyte in solid oxide fuel cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12009-21-1
Molecular Formula BaO3Zr
Molecular Weight (g/mol) 276.548
MDL Number MFCD00014189
InChI Key DQBAOWPVHRWLJC-UHFFFAOYSA-N
Synonym barium zirconate, barium 2+ oxozirconiumbis olate, ba.o3zr, barium 2+ ;dioxido oxo zirconium, barium zirconate, powder, <10 mum, barium zirconate, nanopowder, <50 nm particle size trace metals basis
PubChem CID 16212444
IUPAC Name barium(2+);dioxido(oxo)zirconium
SMILES [O-][Zr](=O)[O-].[Ba+2]

Specifications

Melting Point 2500°C
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99% (Metal basis)
Solubility Information Insoluble in water.
Formula Weight 276.53
Odor Odorless
Appearance Off-white powder
Density 5.52 g/cm3
Chemical Name or Material Barium zirconium oxide
Show More Show Less
Hazard Category H302+H332
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
EINECSNumber 234-546-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.