Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BarX1 Mouse anti-Human, Clone: BARX1/2759, Novus Biologicals™
SDP

Catalog No. NB122878 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB122878 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB122878 Supplier Novus Biologicals Supplier No. NBP30770220UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BarX1 Monoclonal specifically detects BarX1 in Human samples. It is validated for Immunohistochemistry-Paraffin.

Specifications

Antigen BarX1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BARX1/2759
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias BarH-like homeobox 1, BARX homeobox 1, homeobox protein BarH-like 1
Host Species Mouse
Immunogen Recombinant fragment of human BarX1 protein (around aa 155-254) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56033
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.