Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basal cell cytokeratin Antibody (RCK103), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP197729A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP197729A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP197729A70 Supplier Novus Biologicals Supplier No. NBP197729AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Basal cell cytokeratin Monoclonal specifically detects Basal cell cytokeratin in Human, Rat, Porcine, Avian, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Basal cell cytokeratin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RCK103
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen RCK103 is a mouse monoclonal IgG1 antibody derived by fusion of SP2/0-Ag14 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a mix of cell preparations containing Human Cytokeratins.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This monoclonal antibody stains basal cells in combined and stratified epithelial tissues. It recognizes the stem cell population, including the so-called amplifying cells in the prostate epithelium.
Target Species Human, Rat, Pig, Avian, Canine, Chicken, Guinea Pig, Hamster, Rabbit, Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.