Learn More
Eisco™ Basalt Specimen
SCI_ED
Catalog No. p-7076844
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
Basalt is formed quickly from an iron-rich magma and exhibits a dark color and small crystal size.
  • Aphanitic volcanic rock forms from effusive eruptions, such as shield volcanoes of Hawaii, or deep ocean floor
  • For use in introduction to igneous rock classification and identification
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Students can recreate cooling history of this sample based on texture and abundance of phenocrysts
  • Example of aphanitic igneous rock
Specifications
Product Type Igneous Rock, Basalt
For Use With (Application) Introduction to igneous rock classification and identification
Packaging Clear labeled and barcoded bag
Quantity 1 Ea.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.