Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Basophils Antibody (212H6.16), Alexa Fluor™ 546, Novus Biologicals™
SDP

Catalog No. DDX0030A546 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DDX0030A546 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DDX0030A546 Supplier Novus Biologicals Supplier No. DDX0030A546100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Basophils Monoclonal specifically detects Basophils in Canine, Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry.

Specifications

Antigen Basophils
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 212H6.16
Concentration 0.5mg/mL
Conjugate Alexa Fluor 546
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Gene Accession No. NP_006857
Gene Alias Anti-Basophils Basophil
Host Species Mouse
Immunogen Basophil was used as immunogen.
Purification Method Ion exchange chromatography
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Human, Canine, No-cross reactivity on mouse basophil.
Target Species Canine, Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.