Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bassoon Antibody (SAP7F407), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications
Supplier: Novus Biologicals NB120132490.2MG
Description
Bassoon Monoclonal specifically detects Bassoon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Bassoon | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunoprecipitation 2.5 ug, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen | |
O88778 | |
BSN | |
Recombinant rat Bassoon. | |
Protein G purified | |
RUO | |
8927 | |
Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
SAP7F407 | |
Unconjugated | |
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide | |
bassoon (presynaptic cytomatrix protein) | |
Mouse | |
400 kDa | |
0.2 mg | |
Primary | |
This affinity purified detects an ∼400 kDa protein, corresponding to the apparent molecular mass of Bassoon on SDSPAGE immunoblots, in samples from mouse and rat origins. Additional bands between 97 and 400 kDa may also be detected. | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction