Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bassoon Antibody (SAP7F407), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 7 publications
$206.50 - $746.50
Specifications
Antigen | Bassoon |
---|---|
Clone | SAP7F407 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunoprecipitation 2.5 ug, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen |
Applications | Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018066
![]() |
Novus Biologicals
NB120132490.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB12013249
![]() |
Novus Biologicals
NB12013249 |
0.05 mg |
Each for $390.00
|
|
|||||
NB120132498
![]() |
Novus Biologicals
NB120132490.2MG |
0.2 mg |
Each for $746.50
|
|
|||||
Description
Bassoon Monoclonal specifically detects Bassoon in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Bassoon | |
1.0 mg/mL | |
Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Rat, Mouse | |
bassoon (presynaptic cytomatrix protein) | |
BSN | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
400 kDa |
SAP7F407 | |
Western Blot 1:500, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:400, Immunoprecipitation 2.5 ug, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen | |
Monoclonal | |
Purified | |
RUO | |
O88778 | |
8927 | |
Recombinant rat Bassoon. | |
Protein G purified | |
This affinity purified detects an ∼400 kDa protein, corresponding to the apparent molecular mass of Bassoon on SDSPAGE immunoblots, in samples from mouse and rat origins. Additional bands between 97 and 400 kDa may also be detected. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title