Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bat Immunoglobulin Antibody (BT1-4F10), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N223483PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N223483PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N223483PEC5 Supplier Novus Biologicals Supplier No. NBP223483PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bat Immunoglobulin Monoclonal specifically detects Bat Immunoglobulin in Bat samples. It is validated for Flow Cytometry.

Specifications

Antigen Bat Immunoglobulin
Applications Flow Cytometry
Classification Monoclonal
Clone BT1-4F10
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Microchiropteran bat (Eptesicus fuscus) spleen cells.
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Immunoglobulin L (Light) chain.
Target Species Bat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.