Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAT3/BAG6 Rabbit anti-Human, Mouse, Rat, Clone: 8P4D9, Novus Biologicals™
SDP

Catalog No. NB166485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166485 100 μg
NB166486 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166485 Supplier Novus Biologicals Supplier No. NBP315695100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BAT3/BAG6 Monoclonal antibody specifically detects BAT3/BAG6 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen BAT3/BAG6
Applications Western Blot
Classification Monoclonal
Clone 8P4D9
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BAT3HLA-B associated transcript-3, BCL2-associated athanogene 6BAG-6, D6S52EHLA-B-associated transcript 3, G3BAG family molecular chaperone regulator 6, HLA-B associated transcript 3, large proline-rich protein BAT3, Protein G3, Protein Scythe, scythe
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1033-1132 of human BAT3/BAG6 (P46379).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7917
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.