Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BAT3/BAG6 Rabbit anti-Human, Mouse, Rat, Clone: 8P4D9, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | BAT3/BAG6 |
---|---|
Clone | 8P4D9 |
Dilution | Western Blot 1:500 - 1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
BAT3/BAG6 Monoclonal antibody specifically detects BAT3/BAG6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
BAT3/BAG6 | |
Western Blot 1:500 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
BAT3HLA-B associated transcript-3, BCL2-associated athanogene 6BAG-6, D6S52EHLA-B-associated transcript 3, G3BAG family molecular chaperone regulator 6, HLA-B associated transcript 3, large proline-rich protein BAT3, Protein G3, Protein Scythe, scythe | |
A synthetic peptide corresponding to a sequence within amino acids 1033-1132 of human BAT3/BAG6 (P46379). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
8P4D9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Apoptosis, Cancer, Cell Biology | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7917 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title